Ethyl-3,3-diethoxyacrylate ≥95.0%

Leverantör: TCI

Synonyms: 3,3-Diethoxyacrylic Acid Ethyl Ester

D2006-25G D2006-5G
TCIAD2006-25GEA 6750 SEK
TCIAD2006-25G TCIAD2006-5G
Ethyl-3,3-diethoxyacrylate ≥95.0%
Ethyl-3,3-diethoxyacrylate
Formel: C₉H₁₆O₄
Kokpunkt: 132°C
Flampunkt: 102°C
MDL Number: MFCD00012220
CAS nummer: 32002-24-7
EINECS: 250-890-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR