4-(2',6'-Dimethylphenoxy)phthalonitrile ≥99.0%

Leverantör: TCI
Varning

Synonyms: 1,2-Dicyano-4-(2',6'-dimethylphenoxy)benzene

D1983-1G D1983-5G
TCIAD1983-1GEA 990 SEK
TCIAD1983-1G TCIAD1983-5G
4-(2',6'-Dimethylphenoxy)phthalonitrile ≥99.0%
4-(2',6'-Dimethylphenoxy)phthalonitrile
Formel: C₁₆H₁₂N₂O
MDL Number: MFCD00059064
CAS nummer: 221302-75-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 109.0 to 112.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR