3-Decen-2-one ≥93.0%

Leverantör: TCI

D1933-25ML D1933-5ML
TCIAD1933-25MLEA 510 SEK
TCIAD1933-25ML TCIAD1933-5ML
3-Decen-2-one ≥93.0%
3-Decen-2-one
Formel: C₁₀H₁₈O
Kokpunkt: 109°C
Flampunkt: 92°C
MDL Number: MFCD00015700
CAS nummer: 10519-33-2
EINECS: 234-059-0

Order Now

Specifikation testresultat

Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR