9-Decenoic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D1932-25ML
D1932-5ML
TCIAD1932-25MLEA
2350
SEK
TCIAD1932-25ML
TCIAD1932-5ML
9-Decenoic acid ≥98.0%
9-Decenoic acid
Formel:
C₁₀H₁₈O₂ MW: 170,25 g/mol Kokpunkt: 162°C |
MDL Number:
MFCD00036663 CAS nummer: 14436-32-9 EINECS: 238-410-9 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...