Bis(4-nitrophenyl)sulphone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4,4'-Dinitrodiphenyl sulphone, Bis(4-nitrophenyl) sulphone

D1927-5G
TCIAD1927-5GEA 405 SEK
TCIAD1927-5G
Bis(4-nitrophenyl)sulphone ≥98.0%
Bis(4-nitrophenyl)sulphone
Formel: C₁₂H₈N₂O₆S
MW: 308,27 g/mol
MDL Number: MFCD00010447
CAS nummer: 1156-50-9
EINECS: 214-589-9

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR