4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
D1910-5G
D1910-1G
TCIAD1910-5GEA
1750
SEK
TCIAD1910-5G
TCIAD1910-1G
4,4'-Dibromotriphenylamine ≥98.0% (by GC, titration analysis)
4,4'-Dibromotriphenylamine
Formel:
C₁₈H₁₃Br₂N |
MDL Number:
MFCD00060104 CAS nummer: 81090-53-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |