Ethyl-4,4-imethyl-3-oxovalerate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Ethyl 4,4-dimethyl-3-oxopentanoate, Ethyl Pivaloylacetate, 4,4-Dimethyl-3-oxovaleric Acid Ethyl Ester, Pivaloylacetic Acid Ethyl Ester
D1891-25ML D1891-5ML
TCIAD1891-25MLEA 1475 SEK
TCIAD1891-25ML TCIAD1891-5ML
Ethyl-4,4-imethyl-3-oxovalerate ≥98.0%
Ethyl-4,4-imethyl-3-oxovalerate
Formel: C₉H₁₆O₃
MW: 172,22 g/mol
Kokpunkt: 88°C
Flampunkt: 78°C
MDL Number: MFCD00042886
CAS nummer: 17094-34-7
EINECS: 241-162-4
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %