N,N'-Dibenzylethylenediamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: N1,N2-Dibenzylethane-1,2-diamine, DBED

D1807-25ML D1807-500ML
TCIAD1807-25MLEA 1125 SEK
TCIAD1807-25ML TCIAD1807-500ML
N,N'-Dibenzylethylenediamine ≥98.0% (by GC, titration analysis)
N,N'-Dibenzylethylenediamine
Formel: C₁₆H₂₀N₂
MW: 240,35 g/mol
Kokpunkt: 175°C
MDL Number: MFCD00004771
CAS nummer: 140-28-3
EINECS: 205-408-4
UN: 3259
ADR: 8,III
Merck Index: 14,01064

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Freezing point 21.0 to 25.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR