3,3'-Dichloro-4,4'-diisocyanatobiphenyl ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3,3'-Dichlorobiphenyl-4,4'-diyl Diisocyanate
D1541-25G
D1541-5G
TCIAD1541-25GEA
6325
SEK
TCIAD1541-25G
TCIAD1541-5G
3,3'-Dichloro-4,4'-diisocyanatobiphenyl ≥95.0%
3,3'-Dichloro-4,4'-diisocyanatobiphenyl
Formel:
C₁₄H₆Cl₂N₂O₂ MW: 305,11 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00059591 CAS nummer: 5331-87-3 UN: 2811 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Butylamine Method) | min. 95.0 % |
Melting point | 168.0 to 172.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...