Diethylene glycol dimethacrylate ≥97.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Oxybis(ethane-2,1-diyl) bis(2-methylacrylate)

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D1530-25ML D1530-500ML D1530-100ML
TCIAD1530-25MLEA 215 SEK
TCIAD1530-25ML TCIAD1530-500ML TCIAD1530-100ML
Diethylene glycol dimethacrylate ≥97.0% stabilised
Diethylene glycol dimethacrylate
Formel: C₁₂H₁₈O₅
MW: 242,27 g/mol
Kokpunkt: 134°C
Flampunkt: 140°C
MDL Number: MFCD00014931
CAS nummer: 2358-84-1
EINECS: 219-099-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf111f8c5c98e23517171c3a3952bf4980df1527&vid=bf111f8c5c98e23517171c3a3952bf4980df1527&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR