1,3-Diphenylisobenzofuran ≥97.0%
Leverantör: TCI
D1520-25G
D1520-5G
TCIAD1520-25GEA
3275
SEK
TCIAD1520-25G
TCIAD1520-5G
1,3-Diphenylisobenzofuran ≥97.0%
1,3-Diphenylisobenzofuran
Formel:
C₂₀H₁₄O |
MDL Number:
MFCD00005931 CAS nummer: 5471-63-6 EINECS: 226-808-5 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 129.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...