1,2-Dibromotetrafluorobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene

D1479-1G D1479-5G
TCIAD1479-1GEA 495 SEK
TCIAD1479-1G TCIAD1479-5G
1,2-Dibromotetrafluorobenzene ≥98.0%
1,2-Dibromotetrafluorobenzene
Formel: C₆Br₂F₄
MW: 307,87 g/mol
Kokpunkt: 96°C
Smältpunkt: 16°C
Flampunkt: 99°C
MDL Number: MFCD00039209
CAS nummer: 827-08-7
EINECS: 212-564-7

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR