Diosgenin ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Nitogenin, (2'R,4S,5'R,6aR,6bS,8aS,8bR,9S,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol

D1474-1G D1474-10G
TCIAD1474-1GEA 215 SEK
TCIAD1474-1G TCIAD1474-10G
Diosgenin ≥95.0%
Diosgenin
Formel: C₂₇H₄₂O₃
MW: 414,63 g/mol
MDL Number: MFCD00016887
CAS nummer: 512-04-9
EINECS: 208-134-3
Merck Index: 14,03295

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 205.0 to 208.0 deg-C
Specific rotation [a]20/D -123 to -127 deg (C=1, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR