N,N'-Dicarbobenzoxy-L-cystine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (Z-Cystine-OH)₂, (Z-Cys-OH)2, N,N'-Di-Cbz-L-cystine, N,N'-Dibenzyloxycarbonyl-L-cystine

D1197-1G D1197-5G
TCIAD1197-1GEA 215 SEK
TCIAD1197-1G TCIAD1197-5G
N,N'-Dicarbobenzoxy-L-cystine ≥98.0%
N,N'-Dicarbobenzoxy-L-cystine
Formel: C₂₂H₂₄N₂O₈S₂
MW: 508,56 g/mol
MDL Number: MFCD00022030
CAS nummer: 6968-11-2
EINECS: 230-181-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -89.0 to -93.0 deg (C=8.8, AcOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR