2,3-Dibromo-2,3-dimethylbutane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Pinacol Dibromide, sym-Dibromotetramethylethane

D1067-25G D1067-5G
TCIAD1067-25GEA 1875 SEK
TCIAD1067-25G TCIAD1067-5G
2,3-Dibromo-2,3-dimethylbutane ≥98.0%
2,3-Dibromo-2,3-dimethylbutane
Formel: C₆H₁₂Br₂
MW: 243,97 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00053168
CAS nummer: 594-81-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR