Homogentisic acid ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-(2,5-Dihydroxyphenyl)acetic acid, 2,5-Dihydroxyphenylacetic Acid
D1050-1G
D1050-5G
D1050-200MG
TCIAD1050-1GEA
1575
SEK
TCIAD1050-1G
TCIAD1050-5G
TCIAD1050-200MG
Homogentisic acid ≥97.0% (by HPLC, titration analysis)
Homogentisic acid
Formel:
C₈H₈O₄ MW: 168,15 g/mol |
MDL Number:
MFCD00004324 CAS nummer: 451-13-8 EINECS: 207-192-7 Merck Index: 14,04737 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 144.0 to 150.0 deg-C |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...