N-(2,4-Dinitrophenyl)glycine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
N-Dnp-glycine, Dnp-Gly-OH
D1032-1G
D1032-100MG
TCIAD1032-1GEA
965
SEK
TCIAD1032-1G
TCIAD1032-100MG
N-(2,4-Dinitrophenyl)glycine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)glycine
Formel:
C₈H₇N₃O₆ MW: 241,16 g/mol |
MDL Number:
MFCD00024400 CAS nummer: 1084-76-0 EINECS: 214-114-5 UN: 1325 ADR: 4.1,II |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...