Ethyl erucate ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Ethyl docos-13-enoate, Erucic Acid Ethyl Ester, cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate

D1014-25ML D1014-5ML
TCIAD1014-25MLEA 2400 SEK
TCIAD1014-25ML TCIAD1014-5ML
Ethyl erucate ≥95.0%
Ethyl erucate
Formel: C₂₄H₄₆O₂
MW: 366,63 g/mol
Kokpunkt: 233°C
MDL Number: MFCD00027405
CAS nummer: 37910-77-3
EINECS: 253-712-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR