2,3-Dichlorotetrahydrofuran ≥90.0%
Leverantör: TCI
|
Varning
|
D0985-25G
D0985-5G
TCIAD0985-25GEA
4425
SEK
TCIAD0985-25G
TCIAD0985-5G
2,3-Dichlorotetrahydrofuran ≥90.0%
2,3-Dichlorotetrahydrofuran
Formel:
C₄H₆Cl₂O Kokpunkt: 63°C |
MDL Number:
MFCD00059736 CAS nummer: 3511-19-1 EINECS: 222-513-0 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...