Lauroylcholine chloride hydrate ≥98.0%
Leverantör: TCI
Synonyms:
Dodecanoylcholine Chloride
D0973-1G
D0973-5G
TCIAD0973-1GEA
630
SEK
TCIAD0973-1G
TCIAD0973-5G
Lauroylcholine chloride hydrate ≥98.0%
Lauroylcholine chloride hydrate
Formel:
C₁₇H₃₆ClNO₂ |
MDL Number:
MFCD00050185 CAS nummer: 25234-60-0 EINECS: 246-745-7 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Water | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...