Dicinnamalacetone ≥98.0%

Leverantör: TCI

Synonyms: 1,9-Diphenylnona-1,3,6,8-tetraen-5-one, 1,9-Diphenyl-1,3,6,8-nonatetraen-5-one

D0900-10G D0900-25G
TCIAD0900-10GEA 575 SEK
TCIAD0900-10G TCIAD0900-25G
Dicinnamalacetone ≥98.0%
Dicinnamalacetone
Formel: C₂₁H₁₈O
MW: 286.37 g/mol
MDL Number: MFCD00004792
CAS nummer: 622-21-9
EINECS: 210-724-0

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 139.0 to 143.0 deg-C
Solubility in hot Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR