Bis(4-nitrophenyl)sulphide ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4,4'-Dinitrodiphenyl Sulphide, Di(4-nitrophenyl) sulphide
D0847-25G
D0847-5G
TCIAD0847-25GEA
1675
SEK
TCIAD0847-25G
TCIAD0847-5G
Bis(4-nitrophenyl)sulphide ≥99.0%
Bis(4-nitrophenyl)sulphide
Formel:
C₁₂H₈N₂O₄S MW: 276,27 g/mol |
MDL Number:
MFCD00039745 CAS nummer: 1223-31-0 EINECS: 214-950-0 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 159.0 to 162.0 deg-C |
Solubility in hot Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...