Zinc dimethyldithiocarbamate ≥97.0%
Leverantör: TCI
|
Fara
|
D0717-25G
D0717-500G
TCIAD0717-25GEA
215
SEK
TCIAD0717-25G
TCIAD0717-500G
Zinc dimethyldithiocarbamate ≥97.0%
Zinc dimethyldithiocarbamate
Formel:
C₃H₇NS₂ |
MDL Number:
MFCD00064797 CAS nummer: 137-30-4 EINECS: 205-288-3 UN: 2811 ADR: 6.1,II Merck Index: 14,10172 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...