4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
4-(4-Dimethylaminophenylazo)benzenearsonic Acid Hydrochloride
D0659-1G
D0659-25G
TCIAD0659-1GEA
430
SEK
TCIAD0659-1G
TCIAD0659-25G
4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride ≥95.0%
4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride
Formel:
C₁₄H₁₆AsN₃O₃ MW: 385.68 g/mol |
MDL Number:
MFCD00012589 CAS nummer: 73688-85-4 UN: 3465 ADR: 6.1 III |
Specifikation testresultat
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...