N,N-Diethyl-N'-oleoylethylenediamine ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N'-Oleoyl-N,N-diethylethylenediamine

D0517-25G D0517-5G
TCIAD0517-25GEA 3375 SEK
TCIAD0517-25G TCIAD0517-5G
N,N-Diethyl-N'-oleoylethylenediamine ≥97.0%
N,N-Diethyl-N'-oleoylethylenediamine
Formel: C₂₄H₄₈N₂O
MW: 380,66 g/mol
MDL Number: MFCD00027344
CAS nummer: 13282-67-2

Order Now

Specifikation testresultat

Appearance Yellow to Amber to Dark purple clear liquid
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR