1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Diethylene glycol divinyl ether, 1,1'-[Oxybis(ethyleneoxy)]diethene, 1,1'-(Oxybis(ethyleneoxy))diethylene, 1,1'-(Oxybis(ethyleneoxy))diethene, Diethyleneglycoldivinylether, (2-(2-Ethenyloxyethoxy)ethoxy)ethene

Stabilization: Stabilised with potassium hydroxide

D0498-25ML D0498-100ML
TCIAD0498-25MLEA 535 SEK
TCIAD0498-25ML TCIAD0498-100ML
1,1'-[Oxybis(ethyleneoxy)]diethylene ≥98.0% stabilised
1,1'-[Oxybis(ethyleneoxy)]diethylene
Formel: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MW: 158,2 g/mol
Kokpunkt: 197°C
Flampunkt: 71°C
MDL Number: MFCD00054610
CAS nummer: 764-99-8
EINECS: 212-133-3

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR