Dipentylamine mixture ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Diamylamine, Dipentylamine
D0286-25ML
D0286-500ML
TCIAD0286-25MLEA
245
SEK
TCIAD0286-25ML
TCIAD0286-500ML
Dipentylamine mixture ≥97.0% (by GC, titration analysis)
Dipentylamine
Formel:
C₁₀H₂₃N MW: 157,3 g/mol Flampunkt: 51°C |
MDL Number:
MFCD00009499 CAS nummer: 2050-92-2 EINECS: 218-108-3 UN: 1992 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % (total of isomer) |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...