Dipentamethylenethiuram tetrasulphide
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Bis(pentamethylenethiocarbamoyl) Tetrasulphide, Tetrasulphanediylbis(piperidin-1-ylmethanethione)
D0279-25G
D0279-500G
TCIAD0279-25GEA
215
SEK
TCIAD0279-25G
TCIAD0279-500G
Dipentamethylenethiuram tetrasulphide
Dipentamethylenethiuram tetrasulphide
Formel:
C₁₂H₂₀N₂S₆ MW: 384,67 g/mol |
MDL Number:
MFCD00047474 CAS nummer: 120-54-7 EINECS: 204-406-0 |
Specifikation testresultat
Elemental analysis (Sulfuride) | 50.0 to 62.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...