4'-Chlorobiphenyl-4-carboxaldehyde ≥98.0%

Leverantör: TCI EUROPE

Synonyms: [1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde

C3616-1G C3616-5G
TCIAC3616-1GEA 655 SEK
TCIAC3616-1G TCIAC3616-5G
4'-Chlorobiphenyl-4-carboxaldehyde ≥98.0%
4'-Chlorobiphenyl-4-carboxaldehyde
Formel: C₁₃H₉ClO
MW: 216,67 g/mol
MDL Number: MFCD01631911
CAS nummer: 80565-30-6

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR