Chlorocyclohexyldimethylsilane 95,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

C3608-5G C3608-25G
TCIAC3608-5GEA 375 SEK
TCIAC3608-5G TCIAC3608-25G
Chlorocyclohexyldimethylsilane 95,0 GC_ANALYSMETOD
Chlorocyclohexyldimethylsilane
Formel: C₈H₁₇ClSi
MW: 176.76 g/mol
Kokpunkt: 98°C
MDL Number: MFCD00010669
CAS nummer: 71864-47-6
UN: 2987
ADR: 8 II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR