Cefoxitin ≥98.0%

Leverantör: TCI

C3598-1G C3598-5G
TCIAC3598-1GEA 845 SEK
TCIAC3598-1G TCIAC3598-5G
Cefoxitin ≥98.0%
Cefoxitin
Formel: C₁₆H₁₇N₃O₇S₂
Storage Temperature: Frys
MDL Number: MFCD00072014
CAS nummer: 35607-66-0
EINECS: 252-641-2
Merck Index: 14,01936

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR