2-(Chloromethyl)-6-methylpyridine hydrochloride ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
C3596-1G
C3596-5G
TCIAC3596-1GEA
1225
SEK
TCIAC3596-1G
TCIAC3596-5G
2-(Chloromethyl)-6-methylpyridine hydrochloride ≥98.0%
2-(Chloromethyl)-6-methylpyridine hydrochloride
Formel:
C₇H₈ClN·HCl MW: 178,06 g/mol |
MDL Number:
MFCD01691969 CAS nummer: 3099-30-7 EINECS: 221-451-1 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...