Chelerythrine chloride ≥97.0%
Leverantör: TCI
Varning
Synonyms: 1,2-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride
C3584-25MG C3584-100MG
TCIAC3584-25MGEA 1625 SEK
TCIAC3584-25MG TCIAC3584-100MG
Chelerythrine chloride ≥97.0%
Chelerythrine chloride
Formel: C₂₁H₁₈ClNO₄
Smältpunkt: 195°C
Storage Temperature: Frys
MDL Number: MFCD00060717
CAS nummer: 3895-92-9
EINECS: 223-444-9
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Water max. 12.0 %