Carisoprodol ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: Isopropylcarbamic acid 2-[(Carbamoyloxy)methyl]-2-methylpentyl ester, 2-[(Carbamoyloxy)methyl]-2-methylpentyl Isopropylcarbamate

C3573-25G C3573-5G
TCIAC3573-25GEA 2400 SEK
TCIAC3573-25G TCIAC3573-5G
Carisoprodol ≥98.0% (by HPLC, total nitrogen)
Carisoprodol
Formel: C₁₂H₂₄N₂O₄
MW: 260,33 g/mol
Kokpunkt: 170°C
MDL Number: MFCD00057661
CAS nummer: 78-44-4
Merck Index: 14,01842

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR