Ciclopirox olamine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one 2-aminoethanol salt
C3545-1G C3545-5G
TCIAC3545-1GEA 710 SEK
TCIAC3545-1G TCIAC3545-5G
Ciclopirox olamine ≥98.0%
Ciclopirox olamine
Formel: C₁₂H₁₇NO₂·C₂H₇NO
MW: 268,3520 g/mol
Smältpunkt: 143°C
MDL Number: MFCD00078997
CAS nummer: 41621-49-2
EINECS: 255-464-9
Merck Index: 14,02266
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %