7-Chlorothieno[3,2-b]pyridine ≥98.0%
Leverantör: TCI EUROPE
Varning
C3541-1G C3541-5G
TCIAC3541-1GEA 775 SEK
TCIAC3541-1G TCIAC3541-5G
7-Chlorothieno[3,2-b]pyridine ≥98.0%
7-Chlorothieno[3,2-b]pyridine
Formel: C₇H₄ClNS
Storage Temperature: Kylskåp
MDL Number: MFCD08448161
CAS nummer: 69627-03-8
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %