3-Fluoro-4-cyanophenyl-4-ethylbenzoate ≥98.0%
Leverantör: TCI
|
Varning
|
C3540-25G
C3540-5G
TCIAC3540-25GEA
3050
SEK
TCIAC3540-25G
TCIAC3540-5G
3-Fluoro-4-cyanophenyl-4-ethylbenzoate ≥98.0%
3-Fluoro-4-cyanophenyl-4-ethylbenzoate
Formel:
C₁₆H₁₂FNO₂ |
MDL Number:
MFCD00689047 CAS nummer: 86776-50-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...