Cambinol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
CAmbinol, 5-[(2-Hydroxy-1-naphthyl)methyl]-6-phenyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone, 5-[(2-Hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, NSC 112546
C3535-5MG
C3535-25MG
TCIAC3535-5MGEA
2075
SEK
TCIAC3535-5MG
TCIAC3535-25MG
Cambinol ≥98.0%
Cambinol
Formel:
C₂₁H₁₆N₂O₂S MW: 360,43 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD09057543 CAS nummer: 14513-15-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...