6-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-Chloro-6-hydroxybenzaldehyde

C3526-1G C3526-5G
TCIAC3526-1GEA 455 SEK
TCIAC3526-1G TCIAC3526-5G
6-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
6-Chlorosalicylaldehyde
Formel: C₇H₅ClO₂
Storage Temperature: Kylskåp
MDL Number: MFCD01646166
CAS nummer: 18362-30-6

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR