2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Chloro-4,6-di(1-naphthyl)-1,3,5-triazine

C3525-1G C3525-200MG
TCIAC3525-1GEA 3125 SEK
TCIAC3525-1G TCIAC3525-200MG
2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine ≥98.0%
2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine
Formel: C₂₃H₁₄ClN₃
MW: 367,84 g/mol
CAS nummer: 78941-32-9

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR