1-(2-Chloroethoxy)-4-nitrobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

C3523-1G C3523-5G
TCIAC3523-1GEA 695 SEK
TCIAC3523-1G TCIAC3523-5G
1-(2-Chloroethoxy)-4-nitrobenzene ≥98.0%
1-(2-Chloroethoxy)-4-nitrobenzene
Formel: C₈H₈ClNO₃
MW: 201,6100 g/mol
Kokpunkt: 206°C
MDL Number: MFCD00674505
CAS nummer: 3383-72-0

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR