Cerium(III) bis(trifluoromethanesulphonyl)imide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Bis(trifluoromethanesulphonyl)imide Cerium(III) salt, Cerium(III) triflimide

C3516-1G C3516-5G
TCIAC3516-1GEA 1025 SEK
TCIAC3516-1G TCIAC3516-5G
Cerium(III) bis(trifluoromethanesulphonyl)imide ≥98.0%
Cerium(III) bis(trifluoromethanesulphonyl)imide
Formel: C₆CeF₁₈N₃O₁₂S₆
MW: 980,52 g/mol
CAS nummer: 1046099-39-1
UN: 1759
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR