5-Chlorobenzoxazole ≥98.0%

Leverantör: TCI EUROPE
Varning

C3509-5G C3509-25G
TCIAC3509-5GEA 575 SEK
TCIAC3509-5G TCIAC3509-25G
5-Chlorobenzoxazole ≥98.0%
5-Chlorobenzoxazole
Formel: C₇H₄ClNO
MW: 153,57 g/mol
Kokpunkt: 114°C
Storage Temperature: Kylskåp
MDL Number: MFCD00216939
CAS nummer: 17200-29-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR