Chlorprothixene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: (Z)-2-Chloro-9-(3-dimethylaminopropylidene)thioxanthene, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-1-propanamine

C3505-250MG C3505-1G C3505-5G
TCIAC3505-250MGEA 215 SEK
TCIAC3505-250MG TCIAC3505-1G TCIAC3505-5G
Chlorprothixene ≥98.0%
Chlorprothixene
Formel: C₁₈H₁₈ClNS
MW: 315,86 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00869180
CAS nummer: 113-59-7
EINECS: 204-032-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,02188

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR