Curculigoside A ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-[[(2,6-Dimethoxybenzoyl)oxy]methyl]-4-hydroxyphenyl beta-D-Glucopyranoside

C3504-10MG C3504-50MG
TCIAC3504-10MGEA 2650 SEK
TCIAC3504-10MG TCIAC3504-50MG
Curculigoside A ≥98.0%
Curculigoside A
Formel: C₂₂H₂₆O₁₁
MW: 466,44 g/mol
MDL Number: MFCD00800689
CAS nummer: 85643-19-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR