3-Chloro-4-methylbenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3-(5,6,6-Trimethyl-2-norbornyl)-cyclohexanol, isobornylcyclohexanol, 2-Chloro-4-formyltoluene
C3499-1G C3499-5G
TCIAC3499-1GEA 390 SEK
TCIAC3499-1G TCIAC3499-5G
3-Chloro-4-methylbenzaldehyde ≥98.0%
3-Chloro-4-methylbenzaldehyde
Formel: C₈H₇ClO
MW: 154,593 g/mol
Kokpunkt: 232°C
Smältpunkt: 30°C
Storage Temperature: Kylskåp
MDL Number: MFCD06411284
CAS nummer: 3411-03-8
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %