2-Chloroquinoxaline ≥98.0%

Leverantör: TCI EUROPE
Varning

C3492-1G C3492-5G
TCIAC3492-1GEA 510 SEK
TCIAC3492-1G TCIAC3492-5G
2-Chloroquinoxaline ≥98.0%
2-Chloroquinoxaline
Formel: C₈H₅ClN₂
MW: 164,5917 g/mol
Kokpunkt: 100°C
MDL Number: MFCD00043907
CAS nummer: 1448-87-9
EINECS: 215-905-8

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR