Cobalt naphthenate (Co ∼ 8%)

Leverantör: TCI
Fara

Synonyms: Naphthenic Acid Cobalt Salt

C3480-25G C3480-500G
TCIAC3480-25GEA 215 SEK
TCIAC3480-25G TCIAC3480-500G
Cobalt naphthenate (Co ∼ 8%)
Cobalt naphthenate
Formel: C₂₂H₁₄CoO₄
Flampunkt: 48°C
MDL Number: MFCD02094763
CAS nummer: 61789-51-3
EINECS: 263-064-0
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Gray to Dark purple to Black clear liquid to cloudy liquid
Cobalt 7.5 to 8.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR