Cefaclor monohydrate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Fara

Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate

C3478-1G C3478-5G
TCIAC3478-1GEA 1025 SEK
TCIAC3478-1G TCIAC3478-5G
Cefaclor monohydrate ≥98.0% (by HPLC, titration analysis)
Cefaclor monohydrate
Formel: C₁₅H₁₄ClN₃O₄S·H₂O
MW: 367.80 g/mol
Storage Temperature: Frys
MDL Number: MFCD00071999
CAS nummer: 70356-03-5
Merck Index: 14,01912

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +100 to +115 deg (C=0.4, water)
Water 4.5 to 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR