Clemizole hydrochloride ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(4-Chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)benzimidazole hydrochloride
C3468-25MG C3468-100MG
TCIAC3468-25MGEA 4100 SEK
TCIAC3468-25MG TCIAC3468-100MG
Clemizole hydrochloride ≥98.0%
Clemizole hydrochloride
Formel: C₁₉H₂₀ClN₃·HCl
MW: 362,30 g/mol
Smältpunkt: 241°C
Storage Temperature: Kylskåp
MDL Number: MFCD00051435
CAS nummer: 1163-36-6
EINECS: 214-605-4
Merck Index: 14,02346
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6989a2468b4622fbc4bb761d7024c5d54622f146&vid=6989a2468b4622fbc4bb761d7024c5d54622f146&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.00 to 12.00 %